missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3IP Antibody (OTI5F2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73775H
This item is not returnable.
View return policy
Description
RAB3IP Monoclonal specifically detects RAB3IP in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RAB3IP | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| RAB3IP | |
| Full length recombinant protein of human RAB3IP (NP_071901) produced in HEK293T cells. | |
| 0.1 mL | |
| 117177.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5F2 | |
| Western Blot, Immunohistochemistry | |
| FLJ14660, FLJ22548, MGC71495, RAB3A interacting protein (rabin3), rab-3A-interacting protein, Rab3A-interacting protein, rabin-3, RABIN3, SSX2 interacting protein, SSX2-interacting protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction