Learn More
RAB3IP Monoclonal Antibody (OTI5G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809000
Encompass_Preferred
 View more versions of this product
Product Code. 17540824
  • € 540.87 / 100µL
Description
The specific function of this protein remains unknown.
Specifications
Specifications
RAB3IP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96QF0
RAB3IP
Full length human recombit protein of human RAB3IP produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI5G4
Unconjugated
RAB3IP
RABIN3, RABIN8
Mouse
Affinity Chromatography
RUO
117177
-20° C, Avoid Freeze/Thaw Cycles