missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3IP Monoclonal Antibody (OTI5G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809000S
This item is not returnable.
View return policy
Description
RAB3IP Monoclonal antibody specifically detects RAB3IP in Human samples. It is validated for Western BlotSpecifications
| RAB3IP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQ51 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80380 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5G4 | |
| Unconjugated | |
| PDCD1LG2 | |
| B230311A06; gene trap PAT 12; Gtpat12; MGC137223 protein; RAB3A interacting protein; RAB3A interacting protein (rabin3); rab-3A-interacting protein; RAB3A-interacting protein; RAB3IP; rab3ip {ECO:0000250; rabin 3; Rabin3; rabin-3; RABIN8; SSX2 interacting protein; SSX2-interacting protein; UniProtKB:Q96QF0} | |
| Full length human recombinant protein of human PDCD1LG2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction