missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rab4 Recombinant Rabbit Monoclonal Antibody (8I5K1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535427
This item is not returnable.
View return policy
Description
Immunogen sequence: MSQTAMSETY DFLFKFLVIG NAGTGKSCLL HQFIEKKFKD DSNHTIGVEF GSKIINVGGK YVKLQIWDTA GQERFRSVTR SYYRGAAGAL LVYDITSRET.
Protein transport. Probably involved in vesicular traffic.
Specifications
| Rab4 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20338, P56371 | |
| Rab4a | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RAB4A (P20338). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8I5K1 | |
| Unconjugated | |
| Rab4a | |
| AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19341, 5867 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction