missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB40C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86761
This item is not returnable.
View return policy
Description
RAB40C Polyclonal specifically detects RAB40C in Human samples. It is validated for Western Blot.
Specifications
| RAB40C | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| RAB40C, member RAS oncogene family, RAR like, Rar-like protein, RARLRAR (RAS like GTPASE) like, RASL8C, RAS-like GTPase, Ras-like protein family member 8C, RAS-like, family 8, member C, ras-related protein Rab-40C, SOCS box-containing protein RAR3 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human RAB40C | |
| 100 μL | |
| Signal Transduction | |
| 57799 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction