missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB40C Polyclonal Antibody

Product Code. 16610535 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16610535 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16610535 Supplier Invitrogen™ Supplier No. PA5104080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB40C.

RAB40C is a probable substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB40C
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB40C
Gene Accession No. Q8VHQ4, Q96S21
Gene Alias cb453; rab40c; Rab40c, member RAS oncogene family; RAR (RAS like GTPASE) like; RAR like; RAR3; RARL; rar-like protein; RASL8C; RAS-like GTPase; ras-like protein family member 8C; RAS-like, family 8, member C; ras-related protein Rab-40C; SOCS box containing protein RAR3; SOCS box-containing protein RAR3; Unknown (protein for MGC:136966); wu:fk50d06; zgc:136966
Gene Symbols RAB40C
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB40C(Accession Q96S21), corresponding to amino acid residues G246-S281.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224624, 359728, 57799
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.