Learn More
Invitrogen™ EBP50 Recombinant Rabbit Monoclonal Antibody (24GB1270)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Marca:  Invitrogen™ MA556945
 Ver más versiones de este producto
Código de producto. 30276012
  • € 564.00 / 100µL
Este artículo no se puede devolver. Vea la política de devoluciones
Descripción
Descripción
EBP50 Recombinant Monoclonal Antibody for Western Blot, Flow
NHERF1 (Na+/H+ exchanger regulatory factor 1), also known as EBP50 (ezrin, radixin, moesin-binding phosphoprotein 50) is an adaptor protein, which associates with beta-catenin and is required for its localization at the cell-cell junctions, interacts with various G protein-coupled receptors and regulates their traffic, as well as sodium-hydrogen exchange and sodium-dependent phosphate transport. NHERF1/EBP50 inhibits cell motility and is required to suppress anchorage-independent growth. It contains C-terminal ERM (ezrin, radixin, moesin)-binding region and two N-terminal PDZ (postsynaptic-density-95/disc-large/ZO1 homology) domains and is able to form head-to-tail intramolecular conformation to regulate its interactions.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
EBP50
Recombinant Monoclonal
Unconjugated
SLC9A3R1
EBP50; EBP-50; ERM-binding phosphoprotein; ezrin-radixin-moesin binding phosphoprotein-50; ezrin-radixin-moesin-binding phosphoprotein 50; Na(+)/H(+) exchange regulatory cofactor NHE-RF1; Na+/H+ exchange regulatory co-factor; Nherf; NHE-RF; Nherf1; NHERF-1; NPHLOP2; Regulatory cofactor of Na(+)/H(+) exchanger; SLC9A3 regulator 1; Slc9a3r1; So; Sodium-hydrogen exchanger regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1; solute carrier family 9 isoform A3 regulatory factor 1; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 1
Rabbit
Affinity chromatography
RUO
59114, 9368
-20°C
Liquid
Flow Cytometry, Western Blot
24GB1270
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14745, Q9JJ19
SLC9A3R1
A synthesized peptide derived from human EBP50 (aa 51-120).
100 μL
Primary
Human, Rat
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado