Learn More
Invitrogen™ RAB5A Monoclonal Antibody (2E8B11), eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 14-9711-82
Description
Description: This 2E8B11 monoclonal antibody recognizes human Rab5a, which is a GTPase involved in intracellular membrane trafficking. Rab5a is reported to have ubiquitous expression. Applications Reported: The 2E8B11 clone has been reported for use in intracellular flow cytometry, western blot, ELISA, immunocytochemistry of cells and immunohistochemistry of tissue. Applications Tested: This 2E8B11 antibody has been tested by western blot in multiple human cell lines. Applications Tested: This 2E8B11 antibody has been tested by immunocytochemical staining of fixed and permeabilized HeLa cells. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
RAB5 are among the small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB5A is required for the fusion of plasma membranes and early endosomes. Contributes to the regulation of filopodia extension. Required for the exosomal release of SDCBP, CD63, PDCD6IP and syndecan. Regulates maturation of apoptotic cell-containing phagosomes, probably downstream of DYN2 and PIK3C3.
Specifications
| RAB5A | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P20339 | |
| RAB5A | |
| Purified recombinant fragment of human Rab5a (AA: 1-215) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2E8B11 | |
| Unconjugated | |
| RAB5A | |
| 2410015H04Rik; AI663973; AU021172; nnyRab5a; Rab 5; Rab 5A; RAB5; RAB5A; RAB5A, member RAS oncogene family; Ras related protein; RAS-associated protein RAB5A; ras-related protein Rab-5A; Small GTP-binding protein rab5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5868 | |
| 4°C | |
| Liquid | |
| Responsibly packaged |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.