missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB5A Monoclonal Antibody (OTI6D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812808S
This item is not returnable.
View return policy
Description
RAB5A Monoclonal antibody specifically detects RAB5A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RAB5A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| M0RC99, P20339, Q9CQD1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 271457, 5868, 64633 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6D9 | |
| Unconjugated | |
| RAB5A | |
| 2410015H04Rik; AI663973; AU021172; nnyRab5a; Rab 5; Rab 5A; RAB5; RAB5A; RAB5A, member RAS oncogene family; Ras related protein; RAS-associated protein RAB5A; ras-related protein Rab-5A; Small GTP-binding protein rab5 | |
| Full length human recombinant protein of human RAB5A (NP_004153) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction