missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB5A Mouse anti-Human, Clone:2E8B11, Abnova™
Mouse monoclonal antibody raised against recombinant human RAB5A.
Brand: Abnova MAB17298.100ug
This item is not returnable.
View return policy
Specifications
| RAB5A | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-400) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000) Immunofluorescence (1:200-1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by th |
|
| RAB5A | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2E8B11 | |
| RAB5A, member RAS oncogene family | |
| In PBS (0.05% sodium azide) | |
| RAB5 | |
| Recombinant protein corresponding to amino acids 1-215 of of human RAB5A from E. coli. | |
| 5868 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction