Learn More
Rab5b Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99625APC
This item is not returnable.
View return policy
Description
Rab5b Polyclonal antibody specifically detects Rab5b in Human samples. It is validated for Western Blot
Specifications
| Rab5b | |
| Western Blot | |
| RAB5B, member RAS oncogene family, ras-related protein Rab-5B | |
| Endosome Markers, Signal Transduction |
| APC | |
| PBS | |
| 0.1 mL | |
| 5869 |