missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB5C Antibody (SR1405), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22539-50ul
This item is not returnable.
View return policy
Description
RAB5C Monoclonal antibody specifically detects RAB5C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| RAB5C | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1405 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| L1880, MGC117217, RAB5C, member of RAS oncogene family, RAB5C, member RAS oncogene family, RAB5CL, RAB5L, RABLMGC138857, ras-related protein Rab-5C | |
| A synthesized peptide derived from human RAB5C (Uniprot #: P51148) | |
| 50 μg | |
| Endosome Markers, Signal Transduction | |
| 5878 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction