missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB5C Polyclonal Antibody

Product Code. 16569481
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16569481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16569481 Supplier Invitrogen™ Supplier No. PA596646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAGRGGAARP NGPAAGNKIC QFKLVLLGES AVGKSSLVLR FVKGQFHEYQ ESTIGAAFLT QTVCLDDTTV KFEIWDTAGQ ERYHSLAPMY YRGAQAAIVV YDITNTDTFA RAKNWVKELQ RQASPNIVIA LAGNKADLAS KRAVEFQEAQ AYADDNSLLF METSAKTAMN VNEIFMAIAK KLPKNEPQNA TGAPGRNRGV DLQENNPASR SQCCSN; Positive Samples: A-549, BT-474, HL-60, Mouse lung; Cellular Location: Cell membrane, Cytoplasmic side, Early endosome membrane, Lipid-anchor, Melanosome.

The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. The possibility that Rab proteins might also direct the exocytosis from secretory vesicles to the plasma membrane is supported by the observation that in yeast, the SEC4 protein, which is 40% homologous to Rab proteins, is associated with secretory vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB5C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB5C
Gene Accession No. P35278, P51148
Gene Alias AI326010; L1880; RAB; RAB5B; RAB5B, member RAS oncogene family; RAB5C; Rab5c protein; RAB5C, member of RAS oncogene family; RAB5C, member RAS oncogene family; RAB5C, member RAS oncogene family isoform b; RAB5CL; RAB5L; RABL; Ras-related protein Rab-5C; small GTPase; wu:fb98b08; wu:fi16g01
Gene Symbols RAB5C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-216 of human RAB5C (NP_9588421).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19345, 287709, 5878
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.