Learn More
Invitrogen™ RAB6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596368
Description
Immunogen sequence: MSTGGDFGNP LRKFKLVFLG EQSVGKTSLI TRFMYDSFDN TYQATIGIDF LSKTMYLEDR TIRLQLWDTA GQERFRSLIP SYIRDSAAAV VVYDITNVNS FQQTTKWIDD VRTERGSDVI IMLVGNKTDL ADKRQVSIEE GERKAKELNV MFIETSAKAG YNVKQLFRRV AAALPGMEST QDRSREDMID IKLEKPQEQP; Positive Samples: BT-474, OVCAR3, SW480, HT-29, 293T, SH-SY5Y, Mouse brain, Mouse lung, Mouse intestine; Cellular Location: Golgi apparatus, Golgi apparatus membrane, Lipid-anchor.
Rab proteins are also an integral part of endocytic pathways. Rab 6A is a ubiquitously expressed member of the Rab family of proteins and localizes to the Golgi membrane where it regulates retrograde transport from the late endosomes via the Golgi to endoplasmic reticulum (ER) pathway. Three isoforms exist due to alternative splicing events, namely the ubiquitously expressed isoforms Rab 6A' and Rab 6A, and the brain-specific isoform Rab 6B.
Specifications
| RAB6A | |
| Polyclonal | |
| Unconjugated | |
| RAB6A | |
| 2610028L11Rik; AA419671; MNCb-1660; Rab GTPase; Rab6; rab-6; RAB6, member RAS oncogene family; RAB6A; RAB6A, member RAS oncogene family; Rab6b; RAB6B, member RAS oncogene family; ras-related protein Rab-6A; RCO4-3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19346, 5870 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20340, P35279 | |
| RAB6A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human RAB6A (NP_0028602). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.