missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab7a Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98732G
This item is not returnable.
View return policy
Description
Rab7a Polyclonal specifically detects Rab7a in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Rab7a | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| DyLight 488 | |
| CMT2B, FLJ20819, member RAS oncogene family, PSN, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Rab7a. | |
| 0.1 mL | |
| Cellular Markers, Endosome Markers, Neuroscience, Signal Transduction | |
| 7879 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction