missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB8A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550483
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total RAB8A protein.
The protein encoded by this gene is a member of the RAS superfamily which are small GTP/GDP-binding proteins with an average size of 200 amino acids. The RAS-related proteins of the RAB/YPT family may play a role in the transport of proteins from the endoplasmic reticulum to the Golgi and the plasma membrane. This protein shares 97%, 96%, and 51% similarity with the dog RAB8, mouse MEL, and mouse YPT1 proteins, respectively and contains the 4 GTP/GDP-binding sites that are present in all the RAS proteins. The putative effector-binding site of this protein is similar to that of the RAB/YPT proteins. However, this protein contains a C-terminal CAAX motif that is characteristic of many RAS superfamily members but which is not found in YPT1 and the majority of RAB proteins. Although this gene was isolated as a transforming gene from a melanoma cell line, no linkage between MEL and malignant melanoma has been demonstrable. This oncogene is located 800 kb distal to MY09B on chromosome 19p13.1.
Specifications
| RAB8A | |
| Polyclonal | |
| Unconjugated | |
| RAB8A | |
| AA409338; cell line NK14 derived transforming oncogene; MEL; mel transforming oncogene (derived from cell line NK14); mel transforming oncogene (derived from cell line NK14)- RAB8 homolog; mel transforming oncogene (RAB8 homolog); oncogene c-mel; Rab8; Rab8a; RAB8A, member RAS oncogene family; ras-associated protein RAB8; ras-related protein Rab-8A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4218 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P61006 | |
| RAB8A | |
| Synthetic peptide corresponding to a region derived from internal residues of human Ras-related protein Rab-8A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction