missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB9A Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99423AF532
This item is not returnable.
View return policy
Description
RAB9A Polyclonal antibody specifically detects RAB9A in Mouse samples. It is validated for Western Blot
Specifications
| RAB9A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| RAB9A, member RAS oncogene family, RAB9RAB9, member RAS oncogene family, ras-related protein Rab-9A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RAB9A | |
| 0.1 mL | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Signal Transduction | |
| 9367 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction