missing translation for 'onlineSavingsMsg'
Learn More

ABHD12B Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against partial recombinant ABHD12B.

Brand:  Abnova PAB18527.100ug

Product Code. 16053137

  • € 406.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

ABHD12B
Polyclonal
Rabbit polyclonal antibody raised against partial recombinant ABHD12B.
In buffer containing 0.02% sodium azide
BC107142
ABHD12B
Recombinant protein corresponding to amino acids 78-284 of human ABHD12B.
100 μg
Primary
This antibody is specific to ABHD12B.
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Western Blot (1:1000-1:2000) Immunohistochemistry (1:100-1:500) ELISA (1:20000-1:80000) The optimal working dilution should be determined by the end user.
ABHD12B
BEM46L3/C14orf29/MGC129926/MGC129927/c14_5314
Rabbit
Protein A purification
RUO
145447
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only