Learn More
ACACA Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACACA.
Brand: Abnova PAB17282.100ug
Description
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Specifications
| ACACA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ACACA. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q13085 | |
| ACACA | |
| A synthetic peptide corresponding to residues surrounding serine 79 of human ACACA. | |
| RUO | |
| 31 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| ACACA | |
| ACAC/ACC/ACC1/ACCA | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total ACACA protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only