Learn More
ACACA Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACACA.
Brand:  Abnova PAB17278.100ug
Product Code. 16082707
  • € 403.00 / 100µg
Description
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
ACACA
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of ACACA.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Q13085
ACACA
A synthetic peptide corresponding to residues surrounding serine 80 of human ACACA.
RUO
31
Human, Mouse, Rat
Liquid
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user.
ACACA
ACAC/ACC/ACC1/ACCA
Rabbit
100 μg
Primary
This antibody detects endogenous levels of total ACACA Carboxylase protein.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.