Learn More
ACLY Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ACLY.
Brand: Abnova PAB22215.100uL
Description
ATP citrate lyase is the primary enzyme responsible for the synthesis of cytosolic acetyl-CoA in many tissues. The enzyme is a tetramer (relative molecular weight approximately 440,000) of apparently identical subunits. It catalyzes the formation of acetyl-CoA and oxaloacetate from citrate and CoA with a concomitant hydrolysis of ATP to ADP and phosphate. The product, acetyl-CoA, serves several important biosynthetic pathways, including lipogenesis and cholesterogenesis. In nervous tissue, ATP citrate-lyase may be involved in the biosynthesis of acetylcholine. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: TTSAIQNRFKYARVTPDTDWARLLQDHPWLLSQNLVVKPDQLIKRRGKLGLVGVNLTLDGVKSWLKPRLGQEATVGKATGFLKNFLIEPFSpecifications
| ACLY | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ACLY. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P53396 | |
| ACLY | |
| Recombinant protein corresponding to amino acids of human ACLY. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| ACLY | |
| ACL/ATPCL/CLATP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 47 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only