Learn More
ACP1 Rabbit anti-Bovine, Human, Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant ACP1.
Brand: Abnova PAB0152.100ug
Description
The product of this gene belongs to the phosphotyrosine protein phosphatase family of proteins. It functions as an acid phosphatase and a protein tyrosine phosphatase by hydrolyzing protein tyrosine phosphate to protein tyrosine and orthophosphate. This enzyme also hydrolyzes orthophosphoric monoesters to alcohol and orthophosphate. This gene is genetically polymorphic, and three common alleles segregating at the corresponding locus give rise to six phenotypes. Each allele appears to encode at least two electrophoretically different isozymes, Bf and Bs, which are produced in allele-specific ratios. Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Specifications
| ACP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant ACP1. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| HAAP/MGC111030/MGC3499 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Bovine, Human, Pig | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| ACP1 | |
| ACP1 | |
| Recombinant protein corresponding to amino acids 40-80 of human ACP1. | |
| RUO | |
| 52 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only