Learn More
ACSL3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ACSL3.
Brand: Abnova PAB5224.100ug
Description
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme is highly expressed in brain, and preferentially utilizes myristate, arachidonate, and eicosapentaenoate as substrates. The amino acid sequence of this isozyme is 92% identical to that of rat homolog. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| ACSL3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ACSL3. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| ACS3/FACL3/PRO2194 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| ACSL3 | |
| ACSL3 | |
| A synthetic peptide corresponding to N-terminus of human ACSL3. | |
| RUO | |
| 2181 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only