Learn More
Actin-pan, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Actin-pan.
Brand:  Abnova PAB17300.100ug
Product Code. 16022737
  • € 406.00 / 100µg
Specifications
Actin-pan
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of Actin-pan.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Rabbit
100 μg
Primary
Human, Mouse
Liquid
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
P60709;P63261
A synthetic peptide corresponding to residues surrounding tyrosine 53/55 of human Actin-pan.
RUO
This antibody detects endogenous levels of total Actin-pan (alpha/beta/gamma) protein.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.