Learn More
ADCY3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADCY3.
Brand: Abnova PAB19742.100ug
Description
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes. [provided by RefSeq
Specifications
| ADCY3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADCY3. | |
| In serum (0.02% sodium azide) | |
| O60266 | |
| ADCY3 | |
| A synthetic peptide corresponding to 15 amino acids at N-terminus of human ADCY3. | |
| RUO | |
| 109 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:160000) Western Blot (1:500-1000) The optimal working dilution should be determined by the end user. | |
| ADCY3 | |
| AC3/KIAA0511 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only