Learn More
ADCYAP1R1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADCYAP1R1.
Brand: Abnova PAB25587.50ug
Description
This gene encodes type I adenylate cyclase activating polypeptide receptor, which is a membrane-associated protein and shares significant homology with members of the glucagon/secretin receptor family. This receptor mediates diverse biological actions of adenylate cyclase activating polypeptide 1 and is positively coupled to adenylate cyclase. Alternative splicing of two exons of this gene generates four major splice variants, but their full-length nature has not been determined. [provided by RefSeq
Specifications
| ADCYAP1R1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADCYAP1R1. | |
| In PBS (0.09% sodium azide) | |
| PAC1/PACAPR/PACAPRI | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 117 | |
| Human, Monkey | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-7 ug/mL) The optimal working dilution should be determined by the end user. | |
| ADCYAP1R1 | |
| ADCYAP1R1 | |
| A synthetic peptide corresponding to 18 amino acids at N-terminal extracellular domain of human ADCYAP1R1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only