Learn More
ADD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADD1.
Brand: Abnova PAB19805.100ug
Description
Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described. [provided by RefSeq]
Specifications
| ADD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADD1. | |
| In serum (2% BSA, 0.02% sodium azide) | |
| P35611 | |
| ADD1 | |
| A synthetic peptide corresponding to 15 amino acids at C-terminus of human ADD1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000-1:10000) Western Blot (1:200-1000) Immunohistochemistry (1:100-500) The optimal working dilution should be determined by the end user. | |
| ADD1 | |
| ADDA/MGC3339/MGC44427 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 118 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only