missing translation for 'onlineSavingsMsg'
Learn More

Adipoq Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Product Code. 16178990
Change view
Click to view available options
Quantity:
150 μg
Unit Size:
150µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16178990 150 μg 150µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16178990 Supplier Abnova Supplier No. PAB5124.150ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a recombinant Adipoq.

Specifications

Antigen Adipoq
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a recombinant Adipoq.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
Gene Adipoq
Gene Alias 30kDa/APN/Acdc/Acrp30/GBP28/adipo/apM1
Gene Symbols Adipoq
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Adipoq.
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11450
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.