missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipor2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Adipor2.
Brand: Abnova PAB13780.100uL
This item is not returnable.
View return policy
Description
Sequence: NEPAKHRLGCTRTPEPDIRLRKGHQLDDTRGSNNDSpecifications
| Adipor2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Adipor2. | |
| In serum (0.02% sodium azide) | |
| 1110001I14Rik/ADCR2/AI115388/AW554121/D6Ucla1e/Paqr2 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Recognizes mouse Adiponectin Receptor 2. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation | |
| Unconjugated | |
| Immunoprecipitation (1:50) The optimal working dilution should be determined by the end user. | |
| Adipor2 | |
| Adipor2 | |
| A synthetic peptide corresponding to amino acids 2-36 of mouse Adipor2. | |
| RUO | |
| 68465 | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction