Learn More
ADRA1D Rabbit anti-Dog, Guinea pig, Horse, Human, Monkey, Pig, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADRA1D.
Brand: Abnova PAB25550.50ug
Description
Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1D-adrenergic receptor. Similar to alpha-1B-adrenergic receptor gene, this gene comprises 2 exons and a single intron that interrupts the coding region. [provided by RefSeq
Specifications
| ADRA1D | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADRA1D. | |
| In PBS (0.09% sodium azide) | |
| ADRA1/ADRA1A/ADRA1R/ALPHA1/DAR/dJ779E11.2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 146 | |
| Canine, Equine, Guinea Pig, Human, Monkey, Pig | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| ADRA1D | |
| ADRA1D | |
| A synthetic peptide corresponding to 18 amino acids at 2nd extracellular domain of human ADRA1D. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except ADRA1B (56%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |