Learn More
AGTR1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AGTR1.
Brand: Abnova PAB24974.100uL
Description
Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. It acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. This gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. At least five transcript variants have been described for this gene. Additional variants have been described but their full-length nature has not been determined. The entire coding sequence is contained in the terminal exon and is present in all transcript variants. [provided by RefSeq
Specifications
| AGTR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AGTR1. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| AG2S/AGTR1A/AGTR1B/AT1/AT1B/AT1R/AT2R1/AT2R1A/AT2R1B/HAT1R | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 185 | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| AGTR1 | |
| AGTR1 | |
| A synthetic peptide corresponding to amino acids 150-200 of human AGTR1. | |
| 100 μL | |
| Primary | |
| AGTR1 polyclonal antibody detects endogenous levels of AGTR1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only