Learn More
AHR Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AHR.
Brand: Abnova PAB16308.50ug
Description
This gene encodes a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. This receptor has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450. Its ligands included a variety of aromatic hydrocarbons. [provided by RefSeq
Specifications
| AHR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AHR. | |
| In PBS (0.09% sodium azide) | |
| P35869 | |
| AHR | |
| A synthetic peptide (conjugated with KLH) corresponding to human AHR. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6 ug/mL) The optimal working dilution should be determined by the end user. | |
| AHR | |
| bHLHe76 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 196 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only