Learn More
AKAP7, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AKAP7.
Brand: Abnova PAB19833.100ug
Description
This gene encodes a member of the A-kinase anchoring protein (AKAP) family, a group of functionally related proteins that bind to a regulatory subunit (RII) of cAMP-dependent protein kinase A (PKA) and target the enzyme to specific subcellular compartments. AKAPs have a common RII-binding domain, but contain different targeting motifs responsible for directing PKA to distinct intracellular locations. Three alternatively spliced transcript variants encoding different isoforms have been described. Additional variants exist, but their full-length natures have not been determined. [provided by RefSeq
Specifications
| AKAP7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AKAP7. | |
| In serum (0.02% sodium azide) | |
| Q9POM2 | |
| AKAP7 | |
| A synthetic peptide corresponding to 15 amino acids at C-terminus of human AKAP7. | |
| RUO | |
| 9465 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:16000-80000) Western Blot (1:200-500) The optimal working dilution should be determined by the end user. | |
| AKAP7 | |
| AKAP18 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only