Learn More
ALDH1A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ALDH1A1.
Brand: Abnova PAB28574.100uL
Description
This protein belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme. This gene encodes a cytosolic isoform, which has a high affinity for aldehydes. [provided by RefSeq
Sequence: IYDEFVRRSVERAKKYILGNPLTPGVTQGPQIDKEQYDKILDLIESGKKEGAKLECGGGPWGNKGYFVQPTVFSNVTDEMRIAKEEIFGPVQQIMKFKSLDDVIKRANNTFYGLSpecifications
| ALDH1A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ALDH1A1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| P00352 | |
| ALDH1A1 | |
| Recombinant protein corresponding to human ALDH1A1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:2500) The optimal working dilution should be determined by the end user. | |
| ALDH1A1 | |
| ALDC/ALDH-E1/ALDH1/ALDH11/MGC2318/PUMB1/RALDH1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 216 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only