missing translation for 'onlineSavingsMsg'
Learn More

ALDH1B1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16023677
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16023677 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16023677 Supplier Abnova Supplier No. PAB19130.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against full-length recombinant ALDH1B1.

This protein belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. This gene does not contain introns in the coding sequence. The variation of this locus may affect the development of alcohol-related problems. [provided by RefSeq

Specifications

Antigen ALDH1B1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant ALDH1B1.
Dilution Western blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene ALDH1B1
Gene Accession No. P30837
Gene Alias ALDH5/ALDHX/MGC2230
Gene Symbols ALDH1B1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human ALDH1B1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219
Test Specificity It can expression in BGC823 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.