Learn More
ALDOA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ALDOA.
Brand: Abnova PAB2568.400uL
Description
This gene product, Aldolase A (fructose-bisphosphate aldolase) is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Aldolase A is found in the developing embryo and is produced in even greater amounts in adult muscle. Aldolase A expression is repressed in adult liver, kidney and intestine and similar to aldolase C levels in brain and other nervous tissue. Aldolase A deficiency has been associated with myopathy and hemolytic anemia. Alternative splicing of this gene results in multiple transcript variants which encode the same protein. [provided by RefSeq
Specifications
| ALDOA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ALDOA. | |
| In PBS (0.09% sodium azide) | |
| NP_000025;P04075 | |
| ALDOA | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ALDOA. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| ALDOA | |
| ALDA/MGC10942/MGC17716/MGC17767 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 226 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only