missing translation for 'onlineSavingsMsg'
Learn More
Learn More
aldosterone Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against aldosterone.
Brand: Abnova PAB5143.150ug
This item is not returnable.
View return policy
Specifications
| aldosterone | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against aldosterone. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| 4-PREGNEN-11b, 21-DIOL-3, 18, 20-TRIONE 18, 21-DIACETATE 3-CARBOXYMETHYLOXIME. | |
| RUO | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction