Learn More
ALK Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ALK.
Brand: Abnova PAB18082.100ug
Description
The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas (ALCLs). The translocation creates a fusion gene consisting of the ALK (anaplastic lymphoma kinase) gene and the nucleophosmin (NPM) gene: the 3' half of ALK, derived from chromosome 2, is fused to the 5' portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK (leukocyte tyrosine kinase). ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system. [provided by RefSeq]
Specifications
| ALK | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ALK. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| Q9UM73 | |
| ALK | |
| A synthetic peptide corresponding to human ALK. | |
| RUO | |
| 238 | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50∼1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| ALK | |
| CD246/Ki-1/TFG/ALK | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody is specific to ALK. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only