Learn More
AOF2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant AOF2.
Brand: Abnova PAB14107.50ug
Description
This gene encodes a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
| AOF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant AOF2. | |
| In PBS (0.05% sodium azide, 0.05% proclin 300) | |
| O60341 | |
| AOF2 | |
| Recombinant protein corresponding to full length human AOF2. | |
| RUO | |
| 23028 | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| ChIP Assay, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) ChIP (3 ug) The optimal working dilution should be determined by the end user. | |
| AOF2 | |
| BHC110/KDM1/KIAA0601/LSD1 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only