missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3S1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AP3S1.
Brand: Abnova PAB19377.100ug
This item is not returnable.
View return policy
Specifications
| AP3S1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| AP3S1 | |
| CLAPS3/Sigma3A | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 1176 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of AP3S1. | |
| In PBS (0.02% sodium azide) | |
| NP_001275 | |
| AP3S1 | |
| A synthetic peptide corresponding to 19 amino acids near internal region of human AP3S1. | |
| 100 μg | |
| Primary | |
| At least three isoforms of AP3S1 are known to exist; this antibody will detect the two larger isoforms. AP3S1 antibody is predicted to not cross-react with other AP3 protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction