Learn More
APBA1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of APBA1.
Brand:  Abnova PAB25658.100ug
Product Code. 16075827
  • € 619.00 / 100µg
Description
The protein encoded by this gene is a member of the X11 protein family. It is a neuronal adapter protein that interacts with the Alzheimer's disease amyloid precursor protein (APP). It stabilizes APP and inhibits production of proteolytic APP fragments including the A beta peptide that is deposited in the brains of Alzheimer's disease patients. This gene product is believed to be involved in signal transduction processes. It is also regarded as a putative vesicular trafficking protein in the brain that can form a complex with the potential to couple synaptic vesicle exocytosis to neuronal cell adhesion. [provided by RefSeq]
Specifications
Specifications
APBA1
Polyclonal
Unconjugated
Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
APBA1
D9S411E/MINT1/X11/X11A/X11ALPHA
Rabbit
Peptide affinity purification
RUO
320
Human, Mouse, Rat
Liquid
ELISA, Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of APBA1.
In PBS (0.05% sodium azide)
NP_001154
APBA1
A synthetic peptide corresponding to 19 amino acids at N-terminus of human APBA1.
100 μg
Primary
At least two alternatively spliced isoforms are known to exist; this antibody will detect both isoforms. This antibody is predicted to not cross-react with other APBA proteins.
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.