Learn More
AQP9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AQP9.
Brand: Abnova PAB18840.200uL
Description
The aquaporins are a family of water-selective membrane channels. The protein encoded by this gene allows passage of a wide variety of noncharged solutes. It stimulates urea transport and osmotic water permeability; there are contradicting reports about its role in providing glycerol permeability. The encoded protein may also play a role in specialized leukocyte functions such as immunological response and bactericidal activity. [provided by RefSeq
Specifications
| AQP9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AQP9. | |
| In buffer containing 0.02% sodium azide | |
| NM_020980 | |
| AQP9 | |
| A synthetic peptide corresponding to 15 amino acids at N-terminus of human AQP9. | |
| RUO | |
| 366 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:80000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| AQP9 | |
| HsT17287/SSC1 | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only