Learn More
ARFGEF2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ARFGEF2.
Brand: Abnova PAB17638.100ug
Description
ADP-ribosylation factors (ARFs) play an important role in intracellular vesicular trafficking. The protein encoded by this gene is involved in the activation of ARFs by accelerating replacement of bound GDP with GTP and is involved in Golgi transport. It contains a Sec7 domain, which may be responsible for its guanine-nucleotide exchange activity and also brefeldin A inhibition. [provided by RefSeq
Specifications
| ARFGEF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ARFGEF2. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q9Y6D5 | |
| ARFGEF2 | |
| A synthetic peptide corresponding to internal of human ARFGEF2. | |
| RUO | |
| 10564 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
| ARFGEF2 | |
| BIG2/FLJ23723/dJ1164I10.1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total ARFGEF2 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only