Learn More
ARL4C, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ARL4C.
Brand: Abnova PAB22227.100uL
Description
ADP-ribosylation factor-like 4C is a member of the ADP-ribosylation factor family of GTP-binding proteins. ARL4C is closely similar to ARL4A and ARL4D and each has a nuclear localization signal and an unusually high guanine nucleotide exchange rate. This protein may play a role in cholesterol transport. [provided by RefSeq
Sequence: GIIYVVDSVDVDRLEEAKTELHKVTKFAENQGTPLLVIANKQDLPKSLPVAEIEKQLALHELIPATTYHVQPACAIIGEGLTEGMDKLYEMILKRRKSLKQKKKRSpecifications
| ARL4C | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ARL4C. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P56559 | |
| ARL4C | |
| Recombinant protein corresponding to amino acids of human ARL4C. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| ARL4C | |
| ARL7/LAK | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10123 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only