Learn More
ARPC1A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ARPC1A.
Brand: Abnova PAB20270.100uL
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein encoded by gene ARPC1B. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages. [provided by RefSeq
Sequence: HGVSFSASGSRLAWVSHDSTVSVADASKSVQVSTLKTEFLPLLSVSFVSENSVVAAGHDCCPMLFNYDDRGCLTFVSKLDIPKQSIQRNMSAMERFRNMDKRATTEDRNTALETLHQNSITQVSIYEVDKQDCRKFCTTGISpecifications
| ARPC1A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ARPC1A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q92747 | |
| ARPC1A | |
| Recombinant protein corresponding to amino acids of human ARPC1A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| ARPC1A | |
| Arc40/SOP2Hs/SOP2L | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10552 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only