Learn More
ARSI Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ARSI.
Brand: Abnova PAB23040.100uL
Description
Sulfatases (EC 3.1.5.6), such as ARSI, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules (Sardiello et al., 2005 [PubMed 16174644]).[supplied by OMIM
Sequence: TYDNCDGPGVCGFDLHEGENVAWGLSGQYSTMLYAQRASHILASHSPQRPLFLYVAFQAVHTPLQSPREYLYRYRTMGNVASpecifications
| ARSI | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ARSI. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q5FYB1 | |
| ARSI | |
| Recombinant protein corresponding to amino acids of human ARSI. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| ARSI | |
| FLJ16069 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 340075 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only