Learn More
ATG10 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATG10.
Brand: Abnova PAB0732.100uL
Description
Autophagy is a process for the bulk degradation of cytosolic compartments by lysosomes. ATG10 is an E2-like enzyme involved in 2 ubiquitin-like modifications essential for autophagosome formation: ATG12 (MIM 609608)-ATG5 (MIM 604261) conjugation and modification of a soluble form of MAP-LC3 (MAP1LC3A; MIM 601242), a homolog of yeast Apg8, to a membrane-bound form (Nemoto et al., 2003 [PubMed 12890687]).[supplied by OMIM
Specifications
| ATG10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ATG10. | |
| Lyophilized from PBS | |
| Q9H0Y0 | |
| ATG10 | |
| A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to C-terminus of human ATG10. | |
| RUO | |
| 83734 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Immunohistochemistry (1:100-1:1000) Immunofluorescence (1:100-1:1000) The optimal working dilution should be determined by the end user. | |
| ATG10 | |
| APG10/APG10L/DKFZp586I0418/FLJ13954/pp12616 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only