missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG13 (phospho S318), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ATG13.
Brand: Abnova PAB19948.100ug
This item is not returnable.
View return policy
Specifications
| ATG13 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of ATG13. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| KIAA0652 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 9776 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:25000-1:175000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ATG13 | |
| ATG13 | |
| Synthetic phosphopeptide corresponding to residues surrounding S318 of human ATG13. | |
| 100 μg | |
| Primary | |
| The product was affinity purified from monospecific antiserum by immunoaffinity purification. Antiserum was first purified against the phosphorylated form of the immunizing peptide. The resultant affinity purified antibody was then cross adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human ATG13 pS318 protein and the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated human ATG13 is minimal by ELISA and western blot. A BLAST analysis was used to suggest cross reactivity with ATG13 from human based on 100% sequence homology with the immunogen. Reactivity against homologues from other sources is not known. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction