Learn More
ATG9A Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATG9A.
Brand:  Abnova PAB12481.100uL
Product Code. 16099866
  • € 624.00 / 100µL
Specifications
ATG9A
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of ATG9A.
In PBS (30% glycerol, 0.09% sodium azide)
Q7Z3C6
ATG9A
A synthetic peptide corresponding to amino acids 750-839 of human ATG9A.
RUO
79065
Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:250) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
ATG9A
APG9L1/MGD3208
Rabbit
100 μL
Primary
Bovine, Chicken, Human, Mouse, Primate, Rat