missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza Neuraminidase Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Avian Influenza Neuraminidase.
Brand: Abnova PAB13128.100ug
This item is not returnable.
View return policy
Specifications
| Avian Influenza Neuraminidase | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Avian Influenza Neuraminidase. | |
| In PBS (0.02% sodium azide) | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| Unconjugated | |
| ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CAC95053 | |
| A synthetic peptide corresponding to in internal region 16 amino acids of Avian influenza Neuraminidase. | |
| RUO | |
| Influenza virus | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction